Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: ADLEWDAGHK TNVQQCYCYC GGPGDWYLKM LQCCKCKQWF HEACVQCLQK PMLFGDRFYT FICSVCSSGP EYLKRLPLQW VDIAHLCLYN LSVIHKKKYF DSELELMTYI NENWDRLHPG ELADTPKSER YEHVLEALND YKTMEVSNGI EKKGKKKSVG RPPGPYTRKM IQKTAEPLLD KESISENPTL DLPCSIGRTE GTAHSSNTSD VDFTGASSAK ETTSSSISRH YGLSDSRKRT RTGRSWPAAI PHLRRRRGRL PRRALQTQNS EIVKDDEGKE DYQFDELNTE ILNNLADQEL QLNHLKNSIT SYFGAAGRIA CGEKYRVLAR RVTLDGKVQY LVEWEGATAS
Polycomb group (PcG) that specifically binds histone H3 trimethylated at 'Lys-36' (H3K36me3) and recruits the PRC2 complex. Acts by binding to H3K36me3, a mark for transcriptional activation, and recruiting the PRC2 complex, leading to enhance PRC2 H3K27me3 methylation activity. Regulates the transcriptional networks during embryonic stem cell self-renewal and differentiation. Promotes recruitment of the PRC2 complex to the inactive X chromosome in differentiating XX ES cells and PRC2 recruitment to target genes in undifferentiated ES cells. Required to repress Hox genes by enhancing H3K27me3 methylation of the PRC2 complex. In some conditions may act as an inhibitor of PRC2 activity: able to activate the CDKN2A gene and promote cellular senescence by suppressing the catalytic activity of the PRC2 complex locally. Binds to the metal-regulating-element (MRE) of MT1A gene promoter
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hPCl2; Metal regulatory transcription factor 2; metal response element-binding transcription factor 2; Metal-response element DNA-binding protein M96; Metal-response element-binding transcription factor 2; polycomb-like 2; Polycomb-like protein 2; putative DNA binding protein; RP5-976O13.1; tudor domain containing 19A
Gene Aliases: dJ976O13.2; M96; MTF2; PCL2; TDRD19A
UniProt ID: (Human) Q9Y483
Entrez Gene ID: (Human) 22823
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support