Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
The p95 gene (identical to NBS1 and nibrin) is a member of the hMre11/hRad50 double-strand break complex. This protein complex has been implicated in Nijmegen breakage syndrome, an autosomal recessive disorder marked by increased cancer incidence, cell cycle checkpoint deficits, and ionizing radiation sensitivity, thus revealing a direct molecular link between double-strand break repair and cell cycle checkpoint functions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DnaJ (Hsp40) homolog, subfamily C, member 1; DnaJ homolog subfamily C member 1; DnaJ protein homolog MTJ1; DnaJ-like protein; DnaJ-like protein 1
Gene Aliases: 4733401K02Rik; AA960110; D230036H06Rik; DNAJC1; DNAJL1; ERdj1; ERj1p; HTJ1; MTJ1
UniProt ID: (Human) Q96KC8, (Mouse) Q61712
Entrez Gene ID: (Human) 64215, (Mouse) 13418
Molecular Function: chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support