Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MASASTSKY NSHSLENESI KRTSRDGVNR DLTEAVPRLP GETLITDKEV IYICPFNGPI KGRVYITNYR LYLRSLETDS SLILDVPLGV ISRIEKMGGA TSRGENSYGL DITCKDMRNL RFALKQEGHS RRDMFEILTR YAFPLAHSLP LFAFLNEEKF NVDGWTVYNP VEEYRRQGLP NHHWRITFIN KCYELCDTYP ALLVVPYRAS DDDLRRVATF RSRNRIPVLS WIHPENKTVI VRCSQPLVGM SGKRNKDDEK YLDVIRETNK QISKLTIYDA RPSVNAVANK ATGGGYESDD AYHNAELFFL DIHNIHVMRE SLKKVKDIVY PNVEESHWLS SLESTHWLEH IKLVLTGAIQ VAD (1-362 aa encoded by BC030779)
Human MTM1, a 603 amino-acid protein, is mutated in myotubular myopathy. The largely related protein hMTMR2 is found mutated in a recessive form of Charcot-Marie-tooth neuropathy. Myotubularin is primarily a lipid phosphatase that acts on hosphatidylinositol 3-monophosphate and is involved in the regulation of the phosphatidylinositol 3-kinase (PI 3-kinase) pathway and membrane trafficking. Wildtype myotubularin can directly dephosphorylate PI 3-P and PI 4-P in vitro. Thus, it decreases PI 3-P levels by down-regulating PI 3-K activity and by facilitating the degradation of PI 3-P.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Myotubularin; Phosphatidylinositol-3,5-bisphosphate 3-phosphatase; Phosphatidylinositol-3-phosphate phosphatase; X-linked myotubular myopathy gene 1
Gene Aliases: AF073996; CG2; CNM; mKIAA4176; Mtm; MTM1; MTMX; XLMTM
UniProt ID: (Human) Q13496, (Rat) Q6AXQ4, (Mouse) Q9Z2C5
Entrez Gene ID: (Human) 4534, (Rat) 288762, (Mouse) 17772
Molecular Function:
phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support