Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins and Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Product FAQs
  • Documents & Support
  • Manuals & Protocols
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • 快速訂購
            • 登入
              登入
              Don't have an account ? Create Account
              • 帳戶
              • 訂單
              • 訂製產品與專案

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • MTMR12 Antibodies

            Invitrogen

            MTMR12 Polyclonal Antibody

            View all (3) MTMR12 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Tech Support
            Datasheet
            Protocols
            Questions & Answers
            Tech Support

            Cite MTMR12 Polyclonal Antibody

            • Antibody Testing Data (1)
            Application
            MTMR12 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Group 53 Created with Sketch.

            FIGURE: 1 / 1

            MTMR12 Antibody (PA5-62384) in IHC (P)

            Immunohistochemical analysis of MTMR12 in human pancreas using MTMR12 Polyclonal Antibody (Product # PA5-62384) shows strong granular cytoplasmic positivity in exocrine glandular cells.
            View Product

            MTMR12 Antibody in Immunohistochemistry (Paraffin) (IHC (P))

            Product Details

            PA5-62384

            Applications
            Tested Dilution
            Publications

            Immunohistochemistry (Paraffin) (IHC (P))

            1:200-1:500
            -
            Product Specifications

            Species Reactivity

            Human

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            Recombinant protein corresponding to Human MTMR12. Recombinant protein control fragment (Product #RP-101360).
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.1 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7.2, with 40% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2644274

            Product Specific Information

            Immunogen sequence: RPEEIHTNEK EVTEKEVTLH LLPGEQLLCE ASTVLKYVQE DSCQHGVYGR LVCTDFKIAF LGDDESALDN DETQFKNKVI GENDITLHCV DQIYGVFDE

            Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 85%.

            Target Information

            Phosphatidylinositide 3-kinase-derived membrane-anchored phosphatidylinositides, such as phosphatidylinositol 3-phosphate (PtdIns(3)P), regulate diverse cellular processes. The protein encoded by this gene functions as an adaptor subunit in a complex with an active PtdIns(3)P 3-phosphatase. Alternatively spliced transcript variants have been identified, but their biological validity has not been determined.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: 3-PAP; 3-phosphatase adapter protein; 3-phosphatase adapter subunit; Inactive phosphatidylinositol 3-phosphatase 12; Myotubularin-related protein 12; Phosphatidylinositol 3 phosphate 3-phosphatase adapter subunit; phosphatidylinositol-3 phosphate 3-phosphatase adapter subunit; phosphatidylinositol-3 phosphate 3-phosphatase adaptor subunit; phosphatidylinositol-3-phosphate associated protein

            View more View less

            Gene Aliases: 3-PAP; KIAA1682; MTMR12; PIP3AP

            View more View less

            UniProt ID: (Human) Q9C0I1

            View more View less

            Entrez Gene ID: (Human) 54545

            View more View less

            Function(s)

            protein binding phosphatase regulator activity phosphatase

            Process(es)

            regulation of catalytic activity toxin transport
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us About Us
            • Careers Careers
            • Investors Investors
            • News News
            • Social Responsibility Social Responsibility
            • Trademarks
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Policy
            • Price & Freight Policy
            • Cookie偏好

              在此網站使用 Cookie 的選擇

              我們以及我們的附屬公司和供應商使用 cookies 和類似技術來經營我們的網站、識別我們網站的訪客、提供安全登入、收集統計資料以優化網站功能,並提供根據您的興趣量身定制的內容。點擊「全部接受」 以接受所有 cookies,並直接進入網站,或點擊「管理設定」 以查看 cookies 類型的詳細說明,並選擇是否在網站接受某些 cookies。
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            Hong Kong flag icon
            Hong Kong

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-mgmkt:80/100.66.75.22:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0