Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGEEGPPSLE YIQAKDLFPP KELVKEEENL QVPFTVLQGE GVEFLGRAAD ALIAISNYRL HIKFKDSVIN VPLRMIDSVE SRDMFQLHIS CKDSKVVRCH FSTFKQCQEW LSRLSRATAR PAKPEDLFAF AYHAWCLGLT EEDQHTHLCQ PGEHIRCRQE AELARMGFDL QNVWRVSHIN SNYKLCPSYP QKLLVPVWIT DKELENVASF RSWKRIPVVV YRHLRNGAAI ARCSQPEISW WGWRNADDEY LVTSIAKACA LDPGTRATGG SLSTGNNDTG EACDADFDSS LTACSGVEST AAPQKLLILD ARSYTAAVAN RAKGGGCECE EYYPNCEVVF MGMANIHAIR NSFQYLRAVC SQMPDPSNWL SALESTKWLQ HLSVMLKAAV LVANTVDREG RPVLVHCSDG WDRTPQIVAL AKILLDPYYR TLEGFQVLVE SDWLDFGHKF GDRCGHQENV EDQNEQCPVF LQWLDSVHQL LKQFPCLFEF NEAFLVKLVQ HTYSCLYGTF LANNPCEREK RNIYKRTCSV WALLRAGNKN FHNFLYTPSS DMVLHPVCHV RALHLWTAVY LPASSPCTLG EENMDLYLSP VAQSQEFSGR SLDRLPKTRS MDDLLSACDT SSPLTRTSSD PNLNNHCQEV RVGLEPWHSN PEGSETSFVD SGVGGPQQTV GEVGLPPPLP SSQKDYLSNK PFKSHKSCSP SYKLLNTAVP REMKSNTSDP EIKVLEETKG PAPDPSAQDE LGRTLDGIGE PPEHCPETEA VSALSKVISN KCDGVCNFPE SSQNSPTGTP QQAQPDSMLG VPSKCVLDHS LSTVCNPPSA ACQTPLDPST DFLNQDPSGS VASISHQEQL SSVPDLTHGE EDIGKRGNNR NGQLLENPRF GKMPLELVRK PISQSQISEF SFLGSNWDSF QGMVTSFPSG EATPRRLLSY GCCSKRPNSK QMRATGPCFG GQWAQREGVK SPVCSSHSNG HCTGPGGKNQ MWLSSHPKQV SSTKPVPLNC PSPVPPLYLD DDGLPFPTDV IQHRLRQIEA GYKQEVEQLR RQVRELQMRL DIRHCCAPPA EPPMDYEDDF TCLKESDGSD TEDFGSDHSE DCLSEASWEP VDKKETEVTR WVPDHMASHC YNCDCEFWLA KRRHHCRNCG NVFCAGCCHL KLPIPDQQLY DPVLVCNSCY EHIQVSRARE LMSQQLKKPI ATASS
MTMR, a member of the myotubularin family, encodes an intracellular protein of 1, 195 amino acids, and shares approximately 47% sequence identity with MTMR3, spiking to over 70% identity in the catalytic domain and the double zinc finger (FYVE) motif. PCR analysis detects expression of MTMR4 in all human tissues examined except lung, small intestine, stomach, salivary gland, adrenal gland, and uterus. Subcellular localization of MTMR4 in transfected HeLa cells showed a perinuclear distribution, similar to that seen with MTMR3. MTMR4 dephosphorylates para-nitrophenylphosphate and phosphatidylinositol 3-phosphate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp781E194; FYVE domain-containing dual specificity protein phosphatase 2; FYVE-DSP2; MGC163449; MGC163451; MTMR7; Myotubularin-related protein 4; Phosphatidylinositol-3,5-bisphosphate 3-phosphatase; Phosphatidylinositol-3,5-bisphosphate 3-phosphatase MTMR4; Zinc finger FYVE domain-containing protein 11; zinc finger, FYVE domain containing 11
Gene Aliases: FYVE-DSP2; KIAA0647; MTMR4; ZFYVE11
UniProt ID: (Human) Q9NYA4
Entrez Gene ID: (Human) 9110
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support