Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEHIRTTKVE QVKLLDRFST SNKSLTGTLY LTATHLLFID SHQKETWILH HHIASVEKLA LTTSGCPLVI QCKNFRTVHF IVPRERDCHD IYNSLLQLSK QAKYEDLYAF SYNPKQNDSE RLQGWQLIDL AEEYKRMGVP NSHWQLSDAN RDYKICETYP RELYVPRIAS KPIIVGSSKF RSKGRFPVLS YYHQDKEAAI CRCSQPLSGF SARCLEDEHL LQAISKANPV NRYMYVMDTR PKLNAMANRA AGKGYENEDN YSNIRFQFVG IENIHVMRSS LQKLLEVNGT KGLSVNDFYS GLESSGWLRH IKAVMDAAVF LAKAITVENA SVLVHCSDGW DRTSQVCSLG SLLLDSYYRT IKGFMVLIEK DWISFGHKFS ERCGQLDGDP KEVSPVFTQF LECVWHLTEQ FPQAFEFSEA FLLQIHEHIH SCQFGNFLGN CQKEREELKL KEKTYSLWPF LLEDQKKYLN PLYSSESHRF TVLEPNTVSF NFKFWRNMYH QFDRTLHPRQ SVFNIIMNMN EQNKQLEKDI KDLESKIKQR KNKQTDGILT KELLHSVHPE SPNLKTSLCF KEQTLLPVND ALRTIEGSSP ADNRYSEYAE EFSKSEPAVV SLEYGVARMT C
Phosphatase that acts on lipids with a phosphoinositol headgroup (PubMed:19038970, PubMed:22647598). Dephosphorylates phosphatidylinositol 3-phosphate (PtdIns(3)P) and phosphatidylinositol 3,5-bisphosphate (PubMed:19038970, PubMed:22647598) (Probable). Binds with high affinity to phosphatidylinositol 3,5-bisphosphate (PtdIns(3,5)P2) but also to phosphatidylinositol 3-phosphate (PtdIns(3)P), phosphatidylinositol 4-phosphate (PtdIns(4)P), and phosphatidylinositol 5-phosphate (PtdIns(5)P), phosphatidic acid and phosphatidylserine (PubMed:19038970). Negatively regulates ER-Golgi protein transport. Probably in association with MTMR9, plays a role in the late stages of macropinocytosis by dephosphorylating phosphatidylinositol 3-phosphate in membrane ruffles (PubMed:24591580). Acts as a negative regulator of KCNN4/KCa3.1 channel activity in CD4(+) T-cells possibly by decreasing intracellular levels of phosphatidylinositol 3-phosphate (PubMed:15831468). Negatively regulates proliferation of reactivated CD4(+) T-cells (PubMed:16847315). In complex with MTMR9, negatively regulates DNA damage-induced apoptosis (PubMed:19038970, PubMed:22647598). The formation of the MTMR6-MTMR9 complex stabilizes both MTMR6 and MTMR9 protein levels (PubMed:19038970). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Myotubularin-related protein 6; Phosphatidylinositol-3,5-bisphosphate 3-phosphatase MTMR6; Phosphatidylinositol-3-phosphate phosphatase
Gene Aliases: MTMR6
UniProt ID: (Human) Q9Y217
Entrez Gene ID: (Human) 9107
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support