Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MNRHLCVWL FRHPSLNGYL QCHIQLHSHQ FRQIHLDTRL QVFRQNRNCI LHLLSKNWSR RYCHQDTKML WKHKALQKYM ENLSKEYQTL EQCLQHIPVN EENRRSLNRR HAELAPLAAI YQEIQETEQA IEELESMCKK TESCSVAQAG MQ (1-151 aa encoded by BC027731)
Mitochondrial peptide chain release factor that directs the termination of translation in response to the peptide chain non-cognate termination stop codons AGG and AGA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mitochontrial peptide chain release factor 1; MRF-1; mtRF-1; Peptide chain release factor 1, mitochondrial
Gene Aliases: A830062K05Rik; MRF1; MtRF-1; MTRF1; MTTRF1; RF1
UniProt ID: (Human) O75570, (Mouse) Q8K126
Entrez Gene ID: (Human) 9617, (Mouse) 211253
Molecular Function:
translation release factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support