Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MRSRVLWGA ARWLWPRRAV GPARRPLSSG SPPLEELFAR GGPLRTFLER QAGSEAHLKV RRPELLAVIK LLNEKEQELR ETEHLLHDEN EDLRKLAENE ITLCQKEITQ LKHQIILLLV PSEETDENDL ILEVTAGVGG QEAMLFTSEI FDMYQQYAAF KRWHFETLEY FPSELGGLRH ASASIGGSEA YRHMKFEGGV HRVQRVPKTE KQGRVHTSTM TVAILPQPTE INLVINPKDL RIDTKRASGA GGQHVNTTDS AVRIVHLPTG VVSECQQERS QLKNKELAMT KLRAKLYSMH LEEEINKRQN ARKIQIGSKG RSEKIRTYNF PQNRVTDHRI NKTLHDLETF MQGDYLLDEL VQSLKEYADY ESLVEIISQK V (1-380 aa encoded by BC011873)
Mitochondrial peptide chain release factor that directs the termination of translation in response to the peptide chain termination codons UAA and UAG.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mitochondrial release factor 1 like; Mitochondrial translational release factor 1-like; mtRF1a; Peptide chain release factor 1-like, mitochondrial
Gene Aliases: 9130004K12Rik; HMRF1L; MRF1L; MTRF1A; MTRF1L
UniProt ID: (Human) Q96K40, (Mouse) Q8BJU9, (Rat) Q4V7E5
Entrez Gene ID: (Human) 54516, (Mouse) 108853, (Rat) 361473
Molecular Function:
translation release factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support