Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TKKKGATLP QHIPAGCSLQ FIFTWCLEIR AIPKKAFLRA LVDYTSDSAE KRRLQELCSK QGAADYSRFV RDACACLLDL LLAFPSCQPP LSLLLEHLPK LQPRPYSCAS SSLFHPGKLH FVFNIVEFLS TATTEVLRKG VCTGWLALLV ASVLQPNIHA SHEDSGKALA PKISISPRTT NSFHLPDDPS IPIIMVGPGT GIAPFIGFLQ HREKLQEQHP DGNFGAMWLF FGCRHKDRDY LFRKELRHFL KHGILTHLKV SFSRDAPVGE EEAPAKYVQD NIQLHGQQVA RILLQENGHI YVCGDAKNMA KDVHDALVQI ISKEVGVEKL EAMKTLATLK EEKRYLQDIW S (376-725 aa encoded by BC109216)
Methionine is an essential amino acid required for protein synthesis and one-carbon metabolism. Its synthesis is catalyzed by the enzyme methionine synthase. Methionine synthase eventually becomes inactive due to the oxidation of its cob(I)alamin cofactor. The protein encoded by this gene regenerates a functional methionine synthase via reductive methylation. It is a member of the ferredoxin-NADP(+) reductase (FNR) family of electron transferases. Patients of the cbl-E complementation group of disorders of folate/cobalamin metabolism are defective in reductive activation of methionine synthase. Alternative splicing of this gene results in multiple transcript variants encoding distinct isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [methionine synthase]-cobalamin methyltransferase (cob(II)alamin reducing); AqCbl reductase; Aquacobalamin reductase; Methionine synthase reductase; methionine synthase reductase, mitochondrial; MGC129643; MSR
Gene Aliases: cblE; MSR; MTRR
UniProt ID: (Human) Q9UBK8
Entrez Gene ID: (Human) 4552
Molecular Function:
oxidoreductase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support