Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Adding 0.2 mL of distilled water will yield a concentration of 500 µg/mL.
Positive Control - WB: human HEPG2 whole cell, monkey liver tissue, rat liver tissue, rat small intestine tissue. IHC: human liver cancer tissue, rat liver tissue. ICC/IF: HEPG2 cell. Flow: HEPG2 cell.|Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
MTTP encodes the large subunit of the heterodimeric microsomal triglyceride transfer protein. Protein disulfide isomerase (PDI) completes the heterodimeric microsomal triglyceride transfer protein, which has been shown to play a entral role in lipoprotein assembly. Mutations in MTP can cause abetalipoproteinemia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC149819; MGC149820; microsomal triglyceride transfer protein (large polypeptide, 88kDa); Microsomal triglyceride transfer protein large subunit
Gene Aliases: ABL; MTP; MTTP
UniProt ID: (Human) P55157, (Rat) D4A1W8
Entrez Gene ID: (Human) 4547, (Rat) 310900
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support