Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
9-13M1 recognizes the peptide core of gastric mucin M1/MUC5AC), and more specifically with the ‘d’ epitope amongst the a, b, c, d, e, f, g and h protein core epitopes defined by Bara for M1. 9-13M1 and 2-11M1 react exclusively with epitopes located in the the Nterminal cysteine-rich part of the peptide core MUC5AC. MUC5AC is present in primary ovarian mucinous cancer and gastric cancer, but usually absent in colorectal adenocarcinoma, thus showing an expression pattern opposite to MUC2. Anti-MUC5AC may be useful for differential identification of primary mucinous ovarian tumors from colon adenocarcinoma metastatic to the ovary. MUC5AC antibodies may also be useful for identification pancreatic carcinoma and pre-cancerous changes vs. normal pancreas
The antibody 2-25LE reacts with Lewisb blood group antigen, a carbohydrate determinant carried on both glycolipids and glycoproteins, detected on erythrocytes, certain epithelial cells, and in secretions of certain individuals.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gastric mucin; lewis B blood group antigen; Major airway glycoprotein; MUC-5AC; mucin 5, subtypes A and C, tracheobronchial/gastric; mucin 5ac; mucin 5AC, oligomeric mucus/gel-forming pseudogene; Mucin-5 subtype AC, tracheobronchial; Mucin-5AC; TBM; Tracheobronchial mucin
Gene Aliases: 2210005L13Rik; leB; MGM; MUC5; MUC5AC; mucin; TBM
UniProt ID: (Human) Q8WWQ3
Entrez Gene ID: (Human) 4586, (Mouse) 17833
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support