Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MNLEGGGRG GEFGMSAVSC GNGKLRQWLI DQIDSGKYPG LVWENEEKSI FRIPWKHAGK QDYNREEDAA LFKAWALFKG KFREGIDKPD PPTWKTRLRC ALNKSNDFEE LVERSQLDIS DPYKVYRIVP EGAKKGAKQL TLEDPQMSMS HPYTMTTPYP SLPAQQVHNY MMPPLDRSWR DYVPDQPHPE IPYQCPMTFG PRGHHWQGPA CENGCQVTGT FYACAPPESQ APGVPTEPSI RSAEALAFSD CRLHICLYYR EILVKELTTS SPEGCRISHG HTYDASNLDQ VLFPYPEDNG QRKNIEKLLS HLERGVVLWM APDGLYAKRL CQSRIYWDGP LALCNDRPNK LERDQTCKLF DTQQFLSELQ AFAHHGRSLP RFQVTLCFGE EFPDPQRQRK LITAHVEPLL ARQLYYFAQQ NSGHFLRGYD LPEHISNPED YHRSIRHSSI QE (1-451 aa encoded by BC015752)
The IRF (interferon regulatory factor) family of transcription factors are characterized by an unique tryptophan pentad repeat DNA-binding domain. The IRFs are important in the regulation of interferons in response to infection by virus, and in the regulation of interferon-inducible genes. IRF4 is lymphocyte specific and negatively regulates toll-like-receptor (TLR) signaling that is central to the activation of innate and adaptive immune systems. A chromosomal translocation involving this gene and the IgH locus, t(6;14)(p25;q32), may be a cause of multiple myeloma. Alternatively spliced transcript variants have been found.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Interferon regulatory factor 4; IRF-4; LSIRF; Lymphocyte-specific interferon regulatory factor; Multiple myeloma oncogene 1; NF-EM5; PU.1 interaction partner; Sfpi1/PU.1 interaction partner; Transcriptional activator PIP
Gene Aliases: AI385587; IRF-4; IRF4; LSIRF; MUM1; NF-EM5; SHEP8; Spip
UniProt ID: (Human) Q15306, (Mouse) Q64287
Entrez Gene ID: (Human) 3662, (Mouse) 16364
Molecular Function:
winged helix/forkhead transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support