Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSGGGTETPV GCEAAPGGGS KKRDSLGTAG SAHLIIKDLG EIHSRLLDHR PVIQGETRYF VKEFEEKRGL REMRVLENLK NMIHETNEHT LPKCRDTMRD SLSQVLQRLQ AANDSVCRLQ QREQERKKIH SDHLVASEKQ HMLQWDNFMK EQPNKRAEVD EEHRKAMERL KEQYAEMEKD LAKFSTF
This gene encodes a component of BLOC-1 (biogenesis of lysosome-related organelles complex 1). Components of this complex are involved in the biogenesis of organelles such as melanosomes and platelet-dense granules. A mouse model for Hermansky-Pudlak Syndrome is mutated in the murine version of this gene. Alternative splicing results in multiple transcript variants. Read-through transcription exists between this gene and the upstream EEF1E1 (eukaryotic translation elongation factor 1 epsilon 1) gene, as well as with the downstream TXNDC5 (thioredoxin domain containing 5) gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: biogenesis of lysosomal organelles complex-1, subunit 5, muted; Biogenesis of lysosome-related organelles complex 1 subunit 5; BLOC-1 subunit 5; muted homolog; Protein Muted homolog
Gene Aliases: BLOC1S5; BLOS5; MU; MUTED
UniProt ID: (Human) Q8TDH9
Entrez Gene ID: (Human) 63915
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support