Search Thermo Fisher Scientific
It is not recommended to aliquot this antibody.
This antibody is not cross-reactive with other Mad family members at physiological levels.
MAX dimerization protein belongs to a subfamily of MAX-interacting proteins. This protein competes with MYC for binding to MAX to form a sequence-specific DNA-binding complex, acts as a transcriptional repressor (while MYC appears to function as an activator) and is a candidate tumor suppressor.
仅用于科研。不用于诊断过程。未经明确授权不得转售。
蛋白别名: antagonizer of myc transcriptional activity; Max dimerization protein 1; Max dimerizer 1; MAX-binding protein; MGC104659; OTTHUMP00000202211; Protein MAD
基因别名: AW122478; BHLHC58; MAD; Mad-1; MAD1; MXD1
UniProt ID: (Human) Q05195
Entrez Gene ID: (Human) 4084, (Rat) 362391, (Mouse) 17119