Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: KSLVLDNWEK EESGTQLLTE DISDMQSENR FTTSLLMIPL LEIHDNRCNL IPEKQDINST NKTYTLTKKK PNPNTSKVVK LEKNLQSNCE WETVVYGKTE DQLIMTEQAR
A-Myb is expressed at specific times in reproductive tissues, some neural cells, and a subset of normal and neoplastic B lymphocytes. Both A-Myb and B-Myb are expressed in t(14:18) lymphoma cells where they then inhibit cell arrest and apoptotic signaling. Expression of B-Myb rescues cells from p53-induced G1 phase arrest that is mediated by p21, while A-Myb functions as an anti-apoptotic factor by effectively activating the bcl-2 promoter and thereby up-regulating Bcl-2 expression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A-Myb; Myb-like protein 1; Myb-related protein A; MYBA; v-myb avian myeloblastosis viral oncogene homolog-like 1; VMYB
Gene Aliases: A-MYB; AMYB; MYBL1
UniProt ID: (Human) P10243
Entrez Gene ID: (Human) 4603
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support