Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MMEKNTSEGP ACSPEETASE SAKVPTAEPP GEVAVSESTR EEQVPKPQAP APQAPTASTA TKPAPPSEDV PSAPLLLTLD DVSSSSVTVS WEPPERLGRL GLQGYVLELC REGASEWVPV SARPMMVTQQ TVRNLALGDK FLLRVSAVSS AGAGPPAMLD QPIHIRENIE APKIRVPRHL RQTYIRQVGE TVNLQIPFQG KPKPQATWTH NGHALDSQRV SMRTGDQDSI LFIRSAQRSD SGRYELTVRV EDLEAKAVID ILVIEKPGPP SSIRLLDVWG CNAALQWTPP QDTGNTELLG YMVQKADKKT GQWFTVLERY HPTTCTISDL IIGNSYSFRV FSENLCGLST SATVTKELAH IQKADIAAKP KGFIERDFSE APSFTQPLAD HTSTPGYSTQ LFCSVRASPK PKIIWMKNKM EIQGNPKYRA LSEQGVCTLE IRKPSPFDSG VYTCKAINVL GEASVDCRLE VKASAAH
MYBPH binds to myosin; probably involved in interaction with thick myofilaments in the A-band.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H-protein; MyBP-H; Myosin-binding protein H
Gene Aliases: MYBPH
UniProt ID: (Human) Q13203
Entrez Gene ID: (Human) 4608
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support