Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
MYBPH binds to myosin; probably involved in interaction with thick myofilaments in the A-band.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H-protein; MyBP-H; Myosin-binding protein H
Gene Aliases: MYBPH
UniProt ID: (Human) Q13203
Entrez Gene ID: (Human) 4608
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support