Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • MyD88 Antibodies

            OriGene

            MYD88 Monoclonal Antibody (OTI2B2), TrueMAB™

            2 References
            View all (38) MyD88 antibodies

            Promotions

            View available promotion(s)

            Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more

            Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more

            Datasheet
            Protocols
            Questions & Answers
            Catalog # TA502117
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            540.00
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite MYD88 Monoclonal Antibody (OTI2B2), TrueMAB™

            • Antibody Testing Data (10)
            Application
            MYD88 Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 10

            MYD88 Antibody (TA502117) in ICC/IF

            Anti-MYD88 mouse monoclonal antibody (TA502117) immunofluorescent staining of COS7 cells transiently transfected by pCMV6-ENTRY MYD88(RC202253).
            View Product

            MYD88 Antibody in Immunocytochemistry (ICC/IF)
            MYD88 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            MYD88 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            MYD88 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            MYD88 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            MYD88 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            MYD88 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            MYD88 Antibody in Western Blot (WB)
            MYD88 Antibody in Flow Cytometry (Flow)
            MYD88 Antibody in Flow Cytometry (Flow)

            Product Details

            TA502117

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:2,000
            -

            Immunohistochemistry (Paraffin) (IHC (P))

            1:150
            -

            Immunocytochemistry (ICC/IF)

            1:100
            -

            Flow Cytometry (Flow)

            1:100
            -

            Miscellaneous PubMed (Misc)

            -
            View 2 publications 2 publications
            Product Specifications

            Species Reactivity

            Human

            Host/Isotype

            Mouse / IgG2a

            Class

            Monoclonal

            Type

            Antibody

            Clone

            OTI2B2

            Immunogen

            Full length human recombinant protein of human MYD88 produced in HEK293T cell.
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            1 mg/mL

            Purification

            Affinity chromatography

            Storage buffer

            PBS with 1% BSA, 50% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            -20°C, Avoid Freeze/Thaw Cycles

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            Target Information

            Myeloid differentiation Marker 88 (MyD88) is an adaptor molecule for the Toll/IL-1R and is involved in the inflammatory response induced by IL-1, IL-18 and LPS. MyD88 recruits IRAK to IL-1 receptor complex in response to IL-1. This pathway further leads to activation of NF-kB. MyD88 has a wide tissue distribution.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: mutant myeloid differentiation primary response 88; myeloid differentiation primary response gene (88); Myeloid differentiation primary response protein MyD88

            View more View less

            Gene Aliases: MYD88; MYD88D

            View more View less

            UniProt ID: (Human) Q99836

            View more View less

            Entrez Gene ID: (Human) 4615

            View more View less

            Function(s)

            death receptor binding protein binding identical protein binding protein self-association TIR domain binding scaffold/adaptor protein

            Process(es)

            toll-like receptor signaling pathway MyD88-dependent toll-like receptor signaling pathway apoptotic process inflammatory response signal transduction cell surface receptor signaling pathway positive regulation of type I interferon production positive regulation of interleukin-17 production positive regulation of interleukin-23 production positive regulation of interleukin-6 production toll-like receptor 9 signaling pathway negative regulation of apoptotic process positive regulation of I-kappaB kinase/NF-kappaB signaling innate immune response regulation of inflammatory response defense response to Gram-positive bacterium positive regulation of NF-kappaB transcription factor activity response to interleukin-1 3'-UTR-mediated mRNA stabilization cellular response to mechanical stimulus
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-g2k4x:80/100.66.78.186:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0