Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Involved in muscle differentiation (myogenic factor). Induces fibroblasts to differentiate into myoblasts. Probable sequence specific DNA-binding protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHc4; Class C basic helix-loop-helix protein 4; Herculin; Muscle-specific regulatory factor 4; Myf-6; Myogenic factor 6; Myogenic factor 6 (herculin); myogenic factor 6 (herculin); myogenic factor-6
Gene Aliases: BHLHC4; CNM3; herculin; MRF4; MRF4AA; Myf-6; MYF6
UniProt ID: (Human) P23409, (Rat) P19335, (Mouse) P15375
Entrez Gene ID: (Human) 4618, (Rat) 25714, (Mouse) 17878
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support