Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MMMDLFETGS YFFYLDGENV TLQPLEVAEG SPLYPGSDGT LSPCQDQMPP EAGSDSSGEE HVLAPPGLQP PHCPGQCLIW ACKTCKRKSA PTDRRKAAT
Involved in muscle differentiation (myogenic factor). Induces fibroblasts to differentiate into myoblasts. Probable sequence specific DNA-binding protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHc4; Class C basic helix-loop-helix protein 4; Muscle-specific regulatory factor 4; Myf-6; Myogenic factor 6; myogenic factor 6 (herculin); myogenic factor-6
Gene Aliases: BHLHC4; CNM3; MRF4; myf-6; MYF6
UniProt ID: (Human) P23409
Entrez Gene ID: (Human) 4618
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support