Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Cellular myosin that appears to play a role in cytokinesis, cell shape, and specialized functions such as secretion and capping.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MYH14 variant protein; Myosin heavy chain 14; Myosin heavy chain, non-muscle IIc; myosin, heavy polypeptide 14; Myosin-14; NMHC II-C; Non-muscle myosin heavy chain IIc; nonmuscle myosin heavy chain II-C
Gene Aliases: DFNA4; DFNA4A; FP17425; KIAA2034; MHC16; MYH14; MYH17; myosin; NMHC II-C; NMHC-II-C; PNMHH
UniProt ID: (Human) Q7Z406
Entrez Gene ID: (Human) 79784
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support