Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 17
Positive Control: Mouse skeletal muscle tissue lysate, rat skeletal muscle tissue lysate, human skeletal muscle tissue, mouse skeletal muscle tissue, rat skeletal muscle tissue.
Tissue Specificity: Group enriched (skeletal muscle, tongue).
Subcellular Location Cytoplasm.
Sequence Similarities: 100% Mouse/Rat.
Predicted band size: 223 kDa.
MYH2 (Myosin-2) is associated with muscle contraction and is required for cytoskeleton organization.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2A myosin heavy chain; fast 2a myosin heavy chain; inclusion body myopathy 3, autosomal dominant; MyHC-2a; MyHC-IIa; Myosin heavy chain 2; Myosin heavy chain 2a; Myosin heavy chain IIa; myosin heavy chain type IIx; Myosin heavy chain, skeletal muscle, adult 2; myosin, heavy chain 2, skeletal muscle, adult; myosin, heavy polypeptide 2, skeletal muscle, adult; Myosin-2; skeletal muscle myosin heavy chain II-a; type IIA myosin heavy chain
Gene Aliases: IBM3; MHC2A; Myh1; MYH2; MYH2A; MYHas8; MyHC-2A; MyHC-IIA; Myhs-f; Myhs-f1; MYHSA2; Myhsf1; MYPOP
UniProt ID: (Human) Q9UKX2, (Mouse) G3UW82
Entrez Gene ID: (Human) 4620, (Rat) 691644, (Mouse) 17882
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support