Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: KLNGTDPEDV IRNAFACFDE EATGTIQEDY LRELLTTMGD RFTDEEVDEL YREAPIDKKG NFNYIEFTRI LKHGAKDKDD; Positive Samples: LO2, A375, HeLa, K-562, Jurkat
This gene encodes a nonsarcomeric myosin regulatory light chain. This protein is activated by phosphorylation and regulates smooth muscle and non-muscle cell contraction. This protein may also be involved in DNA damage repair by sequestering the transcriptional regulator apoptosis-antagonizing transcription factor (AATF)/Che-1 which functions as a repressor of p53-driven apoptosis. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 8.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Epididymis secretory protein Li 24; HEL-S-24; MLC-2; MLC-2A; MLC-2B; MLC20; Myosin regulatory light chain 12A; Myosin regulatory light chain 12B; Myosin regulatory light chain 2, nonsarcomeric; Myosin regulatory light chain 2-B, smooth muscle isoform; Myosin regulatory light chain 20 kDa; myosin regulatory light chain 3; Myosin regulatory light chain MRLC2; Myosin regulatory light chain MRLC3; Myosin RLC; myosin, light chain 12A, regulatory, non-sarcomeric; myosin, light polypeptide, regulatory, non-sarcomeric (20kD); SHUJUN-1
Gene Aliases: HEL-S-24; MLC-2B; MLCB; MRCL3; MRLC2; MRLC3; MYL12A; MYL12B; MYL2B; MYLC2B; RLC
UniProt ID: (Human) P19105
Entrez Gene ID: (Human) 10627
Molecular Function: actin or actin-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support