Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
Regulatory light chain of myosin. Does not bind calcium.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Myosin light chain 1, embryonic muscle/atrial isoform; Myosin light chain 4; Myosin light chain alkali GT-1 isoform; myosin, atrial/fetal muscle, light chain; myosin, light chain 4, alkali; atrial, embryonic; myosin, light polypeptide 4, alkali; atrial, embryonic
Gene Aliases: ALC1; AMLC; GT1; MLC1; MYL4; PRO1957
UniProt ID: (Human) P12829
Entrez Gene ID: (Human) 4635
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support