Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MLCYVTRPD AVLMEVEVEA KANGEDCLNQ VCRRLGIIEV DYFGLQFTGS KGESLWLNLR NRISQQMDGL APYRLKLRVK FFVEPHLILQ EQTRHIFFLH IKEALLAGHL LCSPEQAVEL SALLAQTKFG DYNQNTAKYN YEELCAKELS SATLNSIVAK HKELEGTSQA SAEYQVLQIV SAMENYGIEW HSVRDSEGQK LLIGVGPEGI SICKDDFSPI NRIAYPVVQM ATQSGKNVYL TVTKESGNSI VLLFKMISTR AASGLYRAIT ETHAFYRCDT VTSAVMMQYS RDLKGHLASL FLNENINLGK KYVFDIKRTS KEVYDHARRA LYNAGVVDLV SRNNQSPSHS PLK (1-352 aa encoded by BC002860)
The ERM protein family members ezrin, radixin, and moesin are cytoskeletal effector proteins linking actin to membrane-bound proteins at the cell surface. Myosin regulatory light chain interacting protein (MYLIP) is a novel ERM-like protein that interacts with myosin regulatory light chain and inhibits neurite outgrowth.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: band 4.1 superfamily member BZF1; cellular modulator of immune recognition (c-MIR); E3 ubiquitin ligase-inducible degrader of the low density lipoprotein receptor; E3 ubiquitin-protein ligase MYLIP; Idol; Inducible degrader of the LDL-receptor; MIR; Myosin regulatory light chain interacting protein; Myosin regulatory light chain-interacting protein; RING-type E3 ubiquitin transferase MYLIP
Gene Aliases: 9430057C20Rik; BM-023; BZF1; I79_001311; IDOL; MIR; MYLIP; PP5242
UniProt ID: (Human) Q8WY64, (Mouse) Q8BM54, (Rat) D3ZDI6
Entrez Gene ID: (Human) 29116, (Chinese hamster) 100760940, (Mouse) 218203, (Rat) 306825
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support