Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Immunogen sequence: SGLSGRKSST GSPTSPLNAE KLESEEDVSQ AFLEAVAEEK PHVKPYFSKT IRDLEVVEGS AARFDCKIEG YPDPEVVWFK DDQSIRESRH FQIDYDEDGN CSLIISDVCG DDDAKYTCKA VNSLGEATCT AELIVETMEE GEGEGEEEEE; Positive Samples: HeLa, HepG2, HT-1080; Cellular Location: Cell projection, Cleavage furrow, Cytoplasm, cytoskeleton, lamellipodium
MYLK (MLCK) is a serine/threonine kinase that phosphorylates myosin regulatory light chains to assist in the production of contractile activity (myosin-actin interaction). MLCK is responsible for smooth muscle contraction via phosphorylation of a specific serine in the N-terminus of myosin light chains (MLC), an event that facilitates myosin interaction with actin filaments. It is a central determinant in the development of vascular permeability and tissue edema formation. In the nervous system it has been shown to control the growth initiation of astrocytic processes in culture and to participate in transmitter release at synapses formed between cultured sympathetic ganglion cells. MLCK acts as a critical participant in signaling sequences that result in fibroblast apoptosis. Smooth muscle and non-muscle isozymes are expressed in a wide variety of adult and fetal tissues and in cultured endothelium with qualitative expression appearing to be neither tissue- nor development-specific. Non-muscle isoform 2 is the dominant splice variant expressed in various tissues. The Telokin isoform, which binds calmodulin, has been found in a wide variety of adult and fetal tissues. MLCK is probably down-regulated by phosphorylation. The protein contains 1 fibronectin type III domain and 9 immunoglobulin-like C2-type domains.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Kinase-related protein; KRP; MLCK; mylk; Myosin light chain kinase, smooth muscle; myosin, light polypeptide kinase; smooth muscle myosin light chain kinase; Telokin
Gene Aliases: 9530072E15Rik; A930019C19Rik; AAT7; AW489456; KRP; MLCK; MLCK1; MLCK108; MLCK210; MSTP083; MYLK; MYLK1; nmMlck; smMLCK
UniProt ID: (Human) Q15746
Entrez Gene ID: (Human) 4638, (Mouse) 107589, (Rat) 288057
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support