Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TLSDQVNRRD FESEKRLRKD LKRTKALLAD AQLMLDHLKN SAPSKREIAQ LKNQLEESEF TCAAAVKARK AMEVEIEDLH LQIDDIAKAK TALEEQLSRL QREKNEIQNR LEEDQEDMNE LMKKHKAAVA QASRDLAQIN DLQAQLEEAN KEKQELQEKL QALQSQVEFL EQSMVDKSLV SRQEAKIREL ETRLEFERTQ VKRLESLASR LKENMEKLTE ERDQRIAAEN REKEQNKRLQ RQLRDTKEEM GELARKEAEA SRKKHELEMD LESLEAANQS LQADLKLAFK RIGDLQAAIE DEMESDENED LINSEGDSDV DSELEDRVDG VKSWLSKNKG PSKAASDDGS LKSSSPTSYW KSLAPDRSDD EHDPLDNTSR PRYSHSYLSD SDTEAKLTET NA
MYO18A may link Golgi membranes to the cytoskeleton and participate in the tensile force required for vesicle budding from the Golgi. Thereby, may play a role in Golgi membrane trafficking and could indirectly give its flattened shape to the Golgi apparatus. Alternatively, in concert with LURAP1 and CDC42BPA/CDC42BPB, has been involved in modulating lamellar actomyosin retrograde flow that is crucial to cell protrusion and migration. May be involved in the maintenance of the stromal cell architectures required for cell to cell contact. Regulates trafficking, expression, and activation of innate immune receptors on macrophages. Plays a role to suppress inflammatory responsiveness of macrophages via a mechanism that modulates CD14 trafficking.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MAJN; Molecule associated with JAK3 N-terminus; myosin 18A; Myosin containing a PDZ domain; myosin containing PDZ domain; myosin-XVIIIa; SP-A receptor subunit SP-R210 alphaS; SP-R210; Surfactant protein receptor SP-R210; Unconventional myosin-XVIIIa
Gene Aliases: CD245; KIAA0216; MYO18A; MYSPDZ; SPR210; TIAF1
UniProt ID: (Human) Q92614
Entrez Gene ID: (Human) 399687
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support