Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MYO1C is a member of the unconventional myosin protein family, which are actin-based molecular motors. The protein is found in the cytoplasm, and one isoform with a unique N-terminus is also found in the nucleus. The nuclear isoform associates with RNA polymerase I and II and functions in transcription initiation. The mouse ortholog of this protein also functions in intracellular vesicle transport to the plasma membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ23903; MMI-beta; Myosin I beta; myosin-I beta; myosin-Ic; nuclear myosin I; Unconventional myosin-Ic
Gene Aliases: MMI-beta; MMIb; MYO1C; myr2; NMI
UniProt ID: (Human) O00159
Entrez Gene ID: (Human) 4641
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support