Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Antibody detects endogenous levels of total MYO5C.
May be involved in transferrin trafficking. Likely to power actin-based membrane trafficking in many physiologically crucial tissues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: myosin 5C; myosin-Vc; Unconventional myosin-Vc
Gene Aliases: 9130003O20Rik; BB085560; C330029I24; MYO5C
UniProt ID: (Human) Q9NQX4
Entrez Gene ID: (Human) 55930, (Rat) 315820, (Mouse) 208943
Molecular Function:
actin binding motor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support