Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: KKKGWWYAHF DGPWIARQME LHPDKPPILL VAGKDDMEMC ELNLEETGLT RKRGAEILPR QFEEIWERCG GIQYLQNAIE SRQARPTYAT AMLQSLLK
This gene encodes a protein involved intracellular vesicle and organelle transport, especially in the hair cell of the inner ear. Mutations in this gene have been found in patients with non-syndromic autosomal dominant and recessive hearing loss.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: myosin-VI; Unconventional myosin-6; Unconventional myosin-VI
Gene Aliases: DFNA22; DFNB37; KIAA0389; MYO6
UniProt ID: (Human) Q9UM54
Entrez Gene ID: (Human) 4646
Molecular Function: actin binding motor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support