Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
This gene encodes a protein involved intracellular vesicle and organelle transport, especially in the hair cell of the inner ear. Mutations in this gene have been found in patients with non-syndromic autosomal dominant and recessive hearing loss.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: myosin VI; myosin-VI; Protein twist; Snell's waltzer; tailchaser; Unconventional myosin-6; Unconventional myosin-VI
Gene Aliases: BC029719; DFNA22; DFNB37; KIAA0389; MYO6; RGD1560646; Sv; Tlc
UniProt ID: (Human) Q9UM54, (Mouse) Q64331
Entrez Gene ID: (Human) 4646, (Mouse) 17920, (Rat) 315840
Molecular Function: actin binding motor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support