Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 12
Antibody detects endogenous levels of total MYOCD.
Smooth muscle cells (SM) and cardiac muscle cells-specific transcriptional factor which uses the canonical single or multiple CArG boxes DNA sequence. Acts as a cofactor of serum response factor (SRF) with the potential to modulate SRF-target genes. Plays a crucial role in cardiogenesis and differentiation of the smooth muscle cell lineage (myogenesis).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Basic SAP coiled-coil transcription activator 2; Myocardin; SRF co-factor protein (cardiac and smooth muscle); SRF cofactor protein; transcription factor myocardin
Gene Aliases: Bsac2; BSAC2A; MYCD; MYOCD; Srfcp
UniProt ID: (Human) Q8IZQ8, (Mouse) Q8VIM5, (Rat) Q8R5I7
Entrez Gene ID: (Human) 93649, (Mouse) 214384, (Rat) 246297
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support