Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MLGKPMNNGL MEKMVEESDE EVCLSSLECL RNQCFDLARK LSETNPQERN PQQNMNIRQH VRPEEDFPGR TPDRNYSDML NLMRLEEQLS PRSRVFASCA KEDGCHERDD
MYT1L, myelin transcription factor 1 family, to which the myelin transcription factor 1-like protein (MYT1L) belongs, is expressed primarily in the developing central nervous system and recruits histone deacetylase (HDAC) to regulate neural transcription. Both MYT1L and the related protein MYT1 interact with SIN3B, a protein that mediates transcriptional repression by binding to HDACs, suggesting that the MYT1 family favor the silencing of genes during neural development. Recent studies suggest that polymorphisms of MYT1L may be associated with schizophrenia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Myelin transcription factor 1-like protein; MyT1-L; neural zinc finger transcription factor 1
Gene Aliases: KIAA1106; MRD39; MYT1L; NZF1; ZC2H2C2; ZC2HC4B
UniProt ID: (Human) Q9UL68
Entrez Gene ID: (Human) 23040
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support