Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
MIP-1gamma is a CC chemokine found in murine blood and a wide variety of murine tissues, with no known human homolog. MIP-1gamma signals through the CCR1 receptor. MIP-1gamma chemoattracts neutrophils, and also inhibits colony formation of bone marrow myeloid immature progenitors. MIP-1gamma contains six cysteines, including the four highly conserved cysteine residues present in CC chemokines.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-C motif chemokine 9; CCF18; chemokine CCL9; Macrophage inflammatory protein 1-gamma; Macrophage inflammatory protein-related protein 2; MIP-1 gamma; MIP-1-gamma; MRP-2; RP23-320E6.3; small inducible cytokine A10; small inducible cytokine A9; Small-inducible cytokine A9
Gene Aliases: CCF18; Ccl9; MRP-2; Mrp2; Scya10; Scya9
UniProt ID: (Mouse) P51670
Entrez Gene ID: (Mouse) 20308, (Rat) 360579
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support