Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
Endotoxin level < 0.01EU/µg by LAL method
Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Magrolimab is an anti-CD47 monoclonal antibody designed to promote the phagocytosis of cancer cells by macrophages. Magrolimab is an anti-CD47 monoclonal antibody. Its purpose is to interfere with the recognition of CD47 by SIRPalpha receptors on macrophages, thereby blocking the self-protection signal used by cancer cells to avoid phagocytosis by macrophages.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hu5F9-G4
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support