Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 7
Immunogen sequence: MLLLPLPLL LFLLCSRAEA GEIIGGTECK PHSRPYMAYL EIVTSNGPSK FCGGFLIRRN FVLTAAHCAG RSITVTLGAH NITEEEDTWQ KLEVIKQFRH PKYNTSTLHH DIMLLKLKEK ASLTLAVGTL PFPSQFNFVP PGRMCRVAGW GRTGVLKPGS DTLQEVKLRL MDPQACSHFR DFDHNLQLCV GNPRKTKSAF KGDSGGPLLC AGVAQGIVSY GRSDAKPPAV FTRISHYRPW INQILQAN (1-247 aa encoded by BC069370)
This gene product is a chymotryptic serine proteinase that belongs to the peptidase family S1. It is expressed in mast cells and thought to function in the degradation of the extracellular matrix, the regulation of submucosal gland secretion, and the generation of vasoactive peptides. In the heart and blood vessels, this protein, rather than angiotensin converting enzyme, is largely responsible for converting angiotensin I to the vasoactive peptide angiotensin II. Angiotensin II has been implicated in blood pressure control and in the pathogenesis of hypertension, cardiac hypertrophy, and heart failure. Thus, this gene product is a target for cardiovascular disease therapies. This gene maps to 14q11. 2 in a cluster of genes encoding other proteases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-chymase; Chymase; chymase 1 preproprotein transcript E; chymase 1 preproprotein transcript I; chymase 1, mast cell; chymase, heart; chymase, mast cell; mast cell; Mast cell chymase 1; Mast cell protease 3; Mast cell protease 5; Mast cell protease I; Mast cell protease III; mMCP-5; rMCP-3; rMCP-5; rMCP-III
Gene Aliases: chymase; CMA1; CYH; CYM; Mcp-5; MCP3P; Mcp5; Mcpt3; Mcpt5; MCT1; MMCP-5
UniProt ID: (Human) P23946, (Rat) P50339, (Mouse) P21844
Entrez Gene ID: (Human) 1215, (Rat) 25627, (Mouse) 17228
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support