Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive Samples: A-549, A-431, HT-29, MCF7, Mouse lung, Mouse small intestine; Cellular Location: Cytoplasmic granule
Immunogen sequence: ITEEEDTWQK LEVIKQFRHP KYNTSTLHHD IMLLKLKEKA SLTLAVGTLP FPSQFNFVPP GRMCRVAGWG RTGVLKPGSD TLQEVKLRLM DPQACSHFRD FDHNLQLCVG NPRKTKSAFK GDSGGPLLCA GVAQGIVSYG RSDAKPPAVF TRISHYRPWI NQILQAN
This gene product is a chymotryptic serine proteinase that belongs to the peptidase family S1. It is expressed in mast cells and thought to function in the degradation of the extracellular matrix, the regulation of submucosal gland secretion, and the generation of vasoactive peptides. In the heart and blood vessels, this protein, rather than angiotensin converting enzyme, is largely responsible for converting angiotensin I to the vasoactive peptide angiotensin II. Angiotensin II has been implicated in blood pressure control and in the pathogenesis of hypertension, cardiac hypertrophy, and heart failure. Thus, this gene product is a target for cardiovascular disease therapies. This gene maps to 14q11. 2 in a cluster of genes encoding other proteases.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-chymase; Chymase; chymase 1 preproprotein transcript E; chymase 1 preproprotein transcript I; chymase 1, mast cell; chymase, heart; chymase, mast cell; mast cell; Mast cell chymase 1; Mast cell protease 5; Mast cell protease I; mMCP-5
Gene Aliases: chymase; CMA1; CYH; CYM; Mcp-5; Mcp5; Mcpt5; MCT1; MMCP-5
UniProt ID: (Human) P23946, (Mouse) P21844
Entrez Gene ID: (Human) 1215, (Mouse) 17228
Molecular Function:
protease
serine protease
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support