Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 5
Positive test controls include: M21, Mouse eye, Rat eye. The target is usually found in the following locations: Endoplasmic reticulum membrane, Golgi apparatus, Melanosome, Single-pass type III membrane protein, trans-Golgi network membrane.
Immunogen sequence: CRRRNGYRAL MDKSLHVGTQ CALTRRCPQE GFDHRDSKVS LQEKNCEPVV PNAPPAYEKL SAEQSPPPYS P
MART1 is involved in melanosome biogenesis by ensuring the stability of GPR143. Plays a vital role in the expression, stability, trafficking, and processing of melanocyte protein PMEL, which is critical to the formation of stage II melanosomes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Antigen LB39-AA; Antigen SK29-AA; MART-1; Melanoma antigen recognized by T-cells 1; melanoma associated antigen recognized by T-cells 1; Protein Melan-A
Gene Aliases: A930034P04Rik; MART-1; MART1; MLANA
UniProt ID: (Human) Q16655
Entrez Gene ID: (Human) 2315, (Mouse) 77836, (Rat) 293890
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support