Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
G-protein coupled receptor for glutamate. Ligand binding causes a conformation change that triggers signaling via guanine nucleotide-binding proteins (G proteins) and modulates the activity of down-stream effectors. Signaling inhibits adenylate cyclase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor GRM4; G protein-coupled receptor GRM6; G protein-coupled receptor, family C, group 1, member D; G protein-coupled receptor, family C, group 1, member F; glutamate receptor, metabotropic 4; glutamate receptor, metabotropic 6; Metabotropic glutamate receptor 4; Metabotropic glutamate receptor 6; mGluR4; mGluR6
Gene Aliases: BC021919; CSNB1B; Gm3; GPRC1D; GPRC1F; GRM4; GRM6; mGlu4; mGlu6; MGLUR4; MGLUR6; nerg1; nob2; nob3; nob4
UniProt ID: (Human) Q14833, (Human) O15303, (Rat) P31423, (Mouse) Q68EF4, (Rat) P35349, (Mouse) Q5NCH9
Entrez Gene ID: (Human) 2914, (Human) 2916, (Rat) 24417, (Mouse) 268934, (Rat) 24419, (Mouse) 108072
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support