Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 8
Immunogen sequence: WTGPKVVSKE IPVESIEEVS KIIKRAPVIW DNIHANDYDQ KRLFLGPYKG RSTELIPRLK GVLTNPNCEF EANYVAIHTL ATWYKSNMNG VRKDVVM
Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
This gene encodes a member of the Ikaros family of zinc-finger proteins. Three members of this protein family (Ikaros, Aiolos and Helios) are hematopoietic-specific transcription factors involved in the regulation of lymphocyte development. This protein forms homo- or hetero-dimers with other Ikaros family members, and is thought to function predominantly in early hematopoietic development. Multiple transcript variants encoding different isoforms have been found for this gene, but the biological validity of some variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-hexosaminidase; Beta-N-acetylglucosaminidase; Beta-N-acetylhexosaminidase; bifunctional protein NCOAT; hyaluronidase in meningioma; Meningioma-expressed antigen 5; N-acetyl-beta-D-glucosaminidase; N-acetyl-beta-glucosaminidase; NCOAT; Nuclear cytoplasmic O-GlcNAcase and acetyltransferase; O-GlcNAcase; OGA; Protein O-GlcNAcase
Gene Aliases: HEXC; KIAA0679; MEA5; MGEA5; NCOAT; OGA
UniProt ID: (Human) O60502
Entrez Gene ID: (Human) 10724
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support