Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: HGLSLIPSTG LCSPDLVNRI IKQEPVLENC SQDLLQHHAD LTCTTTLDLT DGTITFNNNL GTGTEANQAY SVPTKMGSKL EDILMDDTLS PVGVTDPLLS SVSPGASKTS SRRSSMSMEE TEHTC
Mi is a basic helix-loop-helix-leucine zipper (b-HLH-ZIP) transcription factor implicated in pigmentation, mast cells and bone development. The mutation of Mi causes Waardenburg Syndrome type II in humans. In mice, a profound loss of pigmented cells in the skin eye and inner ear results, as well as osteopetrosis and defects in natural killer and mast cells. There are two known isoforms of Mi differing by 66 amino acids at the NH2 terminus. Shorter forms are expressed in melanocytes and run as two bands at 52 kDa and 56 kDa, while the longer Mi form runs as a cluster of bands at 60-70 kDa in osteoclasts and in B16 melonoma cells (but not other melanoma cell lines), as well as mast cells and heart.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHe32; Class E basic helix-loop-helix protein 32; microphtalmia-associated transcription factor; microphthalmia; Microphthalmia-associated transcription factor
Gene Aliases: BHLHE32; CMM8; MI; MITF; WS2; WS2A
UniProt ID: (Human) O75030, (Rat) O88368
Entrez Gene ID: (Human) 4286, (Rat) 25094
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support