Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This protein is implicated in chromosome condensation and DNA damage induced cellular responses, and may play a role in neurogenesis and regulation of the size of the cerebral cortex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BRCT-repeat inhibitor of TERT expression 1; BRIT 1; FLJ12847; Microcephalin; truncated microcephalin
Gene Aliases: 5430437K10Rik; BRIT1; D030046N04Rik; MCPH1; MCT
UniProt ID: (Human) Q8NEM0, (Mouse) Q7TT79
Entrez Gene ID: (Human) 79648, (Mouse) 244329
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support