Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Mitochondria Membrane Integrity cocktail of monoclonal antibodies specific to human, bovine, mouse, rat mitochondria membranes. This antibody is validated for use in Western Blot, and can be used to assess mitochondrial membrane integrity during and after purification.
Contains:
50 µg of Porin clone #2B12AF12 - Human VDAC1. Concentration 2 µg/mL.
50 µg of Cyclophilin D clone #E11AE12BD4 - Rat CypD/PPIF. Concentration 2 µg/mL.
50 µg of C-III-Core 1 clone # 16D10AD9AH5 - Bovine Heart Complex II/III. Concentration 2 µg/mL.
5 µg of Cytochrome C clone #37BA11 - Human Cytochrome c. Concentration 0.1 µg/mL.
100 µg of C-V-a clone #7H10BD4F9 - Bovine Complex V. Concentration 4 µg/mL.
This antibody cocktail (2.37 mg/mL) should be diluted to a final working concentration of 10.2 µg/mL for use in Western blot applications.
Voltage-dependent anion channel (VDAC) proteins are abundant, pore-forming proteins belonging to the eukaryotic mitochondrial porins. It was discovered in the mitochondrial outer membrane. It also is expressed in the plasma membrane. At least three different VDAC genes have been identified in vertebrates. VDAC proteins are known to play an essential role in cellular metabolism and in the early stages of apoptosis. For example, VDAC contitutes a major pathway by which metabolites such as ADP/ATP, succinate and citrate are exchanged between the cytosol and mitochondria. And VDAC1 in the plasma membrane establishes a novel level of apoptosis regulation putatively via its redox activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 16D10AD9AH5; 20B12AF2; 37BA11; 7H10BD4F9; BR1-VDAC; Brain-derived voltage-dependent anion channel 1; Complex III subunit 4; Complex III subunit IV; cyclophilin 3; Cyclophilin D; Cyclophilin F; CyP-D; CyP-M; Cytochrome b-c1 complex subunit 4; Cytochrome c; Cytochrome C, expressed in somatic tissues; Cytochrome c, somatic; Cytochrome c-1; Cytochrome c1, heme protein, mitochondrial; E11AE12BD4; Mitochondrial cyclophilin; mitochondrial Cyclophilin D; mVDAC1; mVDAC5; Non-selective voltage-gated ion channel VDAC1; Outer mitochondrial membrane protein porin 1; Peptidyl-prolyl cis-trans isomerase F, mitochondrial; peptidyl-prolyl cis-trans isomerase, mitochondrial; peptidylprolyl isomerase F; Plasmalemmal porin; Porin 31HL; Porin 31HM; PPIase F; Rotamase F; rVDAC1; Ubiquinol-cytochrome-c reductase complex cytochrome c1 subunit; VDAC-1; VDAC-5; Voltage-dependent anion-selective channel protein 1; Voltage-dependent anion-selective channel protein 5
Gene Aliases: 2610002H19Rik; AA408921; AL033343; AW457192; CYC; CYC1; CYCS; CYCSA; CyP-D; CyP-F; CyP-M; CYP3; CypD; cytc; HCS; MC3DN6; PORIN; PPIase; PPIF; THC4; UQCR4; VDAC; VDAC-1; VDAC1; Vdac5
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support