Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration.
Purity: The precipitin lines against goat anti-mouse whole serum are the same as those against goat anti-mouse IgG, Fc fragment specific when assayed by immunoelectrophoresis at an antigen concentration of 20 mg/mL. There was no reaction against goat anti-mouse IgG, F(ab')2 fragment specific.
This antibody is recommended for use in blocking assays, immunohistochemical, immunocytochemical and immunofluorescent applications.
Storage: After opening, dilute only enough antibody for a single day’s use.
Country of Origin: USA
The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody's species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support