Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Description: The monoclonal mouse IgG1 K immunoglobulin is useful as an isotype control.
Applications Reported: The PerCP-Cy5.5 Mouse IgG1 K Isotype Control has been reported for use in flow cytometric analysis.
Applications Tested: The Mouse IgG1 K Isotype Control has been tested by flow cytometric analysis. Use isotype control at the same concentration as experimental antibody.
Excitation: 488 nm; Emission: 695 nm; Laser: Blue Laser.
Filtration: 0.2 µm post-manufacturing filtered.
Mouse IgG1 is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG1 is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG1 is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Immunoglobulin G; Immunoglobulin G1; ImmunoglobulinG; ImmunoglobulinG1
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support