Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Osenses
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: Mucolipin 2.
Mucolipins constitute a family of cation channel proteins with homologs in mouse, Drosophila, and C. elegans. Mutations in the human MCOLN1 gene cause mucolipodosis IV.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ36691; MCLN2; Mucolipin-2; Transient receptor potential channel mucolipin 2; TRPML2
Gene Aliases: MCOLN2; TRP-ML2; TRPML2
UniProt ID: (Human) Q8IZK6
Entrez Gene ID: (Human) 255231
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support