Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Myeloid differentiation Marker 88 (MyD88) is an adaptor molecule for the Toll/IL-1R and is involved in the inflammatory response induced by IL-1, IL-18 and LPS. MyD88 recruits IRAK to IL-1 receptor complex in response to IL-1. This pathway further leads to activation of NF-kB. MyD88 has a wide tissue distribution.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mutant myeloid differentiation primary response 88; myeloid differentiation primary response 88; myeloid differentiation primary response gene (88); myeloid differentiation primary response protein 88; Myeloid differentiation primary response protein MyD88
Gene Aliases: MYD88; MYD88D
UniProt ID: (Human) Q99836, (Rhesus monkey) B3Y683, (Mouse) P22366
Entrez Gene ID: (Human) 4615, (Rhesus monkey) 696494, (Mouse) 17874
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support