Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TMP21 is a ubiquitously expressed protein that is involved in vesicular targeting and protein transport. More recent experiments have shown that it is also a component in the presenilin complex and modulates the -gamma-secretase but not the epsilon-secretase cleavage activity of the amyloid precursor protein. The presenilin complex is composed of the proteins APH1, nicastrin, and PEN2 in addition to presenilin-1. Together, these proteins cleave the amyloid precursor protein at what is known as the -gamma- and epsilon-sites and can lead to the accumulation of the Abeta cleavage product that is associated with Alzheimer's disease. Co-immunoprecipitation experiments using antibodies against these proteins also yielded TMP21 indicating that TMP21 may play a role in the regulation of this complex. Suppression of TMP21 expression by siRNA in transfected cells caused increased -gamma-secretase activity but not epsilon-secretase activity, and increased Abeta production, demonstrating that TMP21 can modulate -gamma-secretase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cellular myosin heavy chain, type A; flectin; MGC104539; Myosin heavy chain 9; myosin heavy chain IX; Myosin heavy chain, non-muscle IIa; myosin IIA; myosin, heavy polypeptide 9, non-muscle; Myosin-9; myosin9; NMHC II-A; NMHC-II A; NMMHC II-a; NMMHC IIA; NMMHC-A; NMMHC-IIA; non-muscle myosin heavy chain 9; Non-muscle myosin heavy chain A; Non-muscle myosin heavy chain IIa; non-muscle myosin heavy polypeptide 9; nonmuscle myosin heavy chain II-A
Gene Aliases: BDPLT6; DFNA17; EPSTS; Fltn; FTNS; MHA; MYH9; Myhn-1; Myhn1; NMHC-II-A; NMHCIIA; NMMHC-A; NMMHC-IIA; NMMHCA; TU72.6
UniProt ID: (Human) P35579, (Dog) Q258K2, (Mouse) Q8VDD5
Entrez Gene ID: (Human) 4627, (Dog) 481280, (Mouse) 17886
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support