Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Keep as concentrated solution.
Predicted reactivity: Mouse (95%), Rat (94%), Pig (96%), Rabbit (94%), Sheep (94%), Bovine (94%).
Positive Control: RD.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Myogenin is a muscle-specific transcription factor that can induce myogenesis in a variety of cell types in tissue culture. It is a member of a large family of proteins related by sequence homology, the helix-loop-helix proteins. It is essential for the development of functional skeletal muscle.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHc3; Class C basic helix-loop-helix protein 3; Myf-4; Myogenic factor 4; Myogenic factor-4; Myogenin
Gene Aliases: BHLHC3; myf-4; MYF4; MYOG
UniProt ID: (Human) P15173
Entrez Gene ID: (Human) 4656
Molecular Function: basic helix-loop-helix transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support