Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody well recognized endogenous myoglobin in muscle lysates.
Immunogen sequence: MGLSDGEWQ LVLNVWGKVE ADIPGHGQEV LIRLFKGHPE TLEKFDKFKH LKSEDEMKAS EDLKKHGATV LTALGGILKK KGHHEAEIKP LAQSHATKHK IPVKYLEFIS ECIIQVLQSK HPGDFGADAQ GAMNKALELF RKDMASNYKE LGFQG (1-154 aa encoded by BC014547)
This gene encodes a member of the globin superfamily and is expressed in skeletal and cardiac muscles. The encoded protein is a hemoprotein contributing to intracellular oxygen storage and transcellular facilitated diffusion of oxygen. At least three alternatively spliced transcript variants encoding the same protein have been reported.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Human myoglobin; MGC13548; Myoglobin; Nitrite reductase MB; OTTHUMP00000197936; Pseudoperoxidase MB
Gene Aliases: AI325109; MB; myoglobgin; PVALB
UniProt ID: (Human) P02144, (Mouse) P04247, (Rat) Q9QZ76
Entrez Gene ID: (Human) 4151, (Mouse) 17189, (Rat) 59108
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support